missing translation for 'onlineSavingsMsg'
Learn More

Sirtinol, R&D Systems™

Product Code. 18033822
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18033822 50 mg 50mg
18004632 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18033822 Supplier Tocris Bioscience Supplier No. 3521/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective sirtuin family deacetylase inhibitor

Cell-permeable, selective sirtuin deacetylase inhibitor (IC50 values are 38, 68 and 131 μM at SIRT2, Sir2p and SIRT1 respectively) that has no effect on HDAC1 activity. Significantly decreases growth and viability of PCa and HEK293T cells in vitro.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 410536-97-9
Molecular Formula C26H22N2O2
Molecular Weight (g/mol) 394.47
MDL Number MFCD00810186
InChI Key YUGODMKHHCZZOI-ZVTCDHROSA-N
Synonym 2-e-2-hydroxynaphthalen-1-yl methylidene amino-n-1-phenylethyl benzamide
PubChem CID 19577289
IUPAC Name 2-({[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]methyl}amino)-N-[(1S)-1-phenylethyl]benzamide
SMILES C[C@H](NC(=O)C1=CC=CC=C1N\C=C1/C(=O)C=CC2=CC=CC=C12)C1=CC=CC=C1

Especificaciones

Quantity 50 mg
Formula Weight 394.47
Percent Purity >98%
Product Line Tocris
Chemical Name or Material Sirtinol
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.