missing translation for 'onlineSavingsMsg'
Learn More

Sirtinol, MedChemExpress

Product Code. 16498407
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498407 5 mg 5mg
16495718 10 mg 10mg
16421887 50 mg 50mg
16406216 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498407 Supplier Medchem Express Supplier No. HY13515
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sirtinol is a sirtuin (SIRT) inhibitor, with IC50s of 48 μM, 57.7 μM and 131 μM for ySir2, hSIRT2 and hSIRT2, respectively.

Sirtinol is a sirtuin (SIRT) inhibitor, with IC50s of 48 μM, 57.7 μM and 131 μM for ySir2, hSIRT2 and hSIRT2, respectively.

Specifications

Chemical Name or Material Sirtinol
CAS 410536-97-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C26H22N2O2
Quantity 5 mg
Solubility Information DMSO : 10 mg/mL (25.35 mM; Need ultrasonic)
SMILES O=C(NC(C1=CC=CC=C1)C)C2=CC=CC=C2/N=C/C3=C4C=CC=CC4=CC=C3O
Molecular Weight (g/mol) 394.47
Formula Weight 394.47
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.