missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIRT7 Monoclonal Antibody (1E2B2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531903
This item is not returnable.
View return policy
Description
MA5-31903 has been tested in indirect ELISA.
SIRT7 is a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. SIRT7 is included in class IV. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. SIRT7 is included in class IV of the sirtuin family.
Specifications
| SIRT7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q9NRC8 | |
| SIRT7 | |
| Purified recombinant fragment of human SIRT7 (AA: 1-105) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 1E2B2 | |
| Unconjugated | |
| SIRT7 | |
| MGC126840; MGC126842; NAD-dependent deacetylase sirtuin-7; NAD-dependent protein deacetylase sirtuin-7; NAD-dependent protein deacylase sirtuin-7; regulatory protein SIR2 homolog 7; silent mating type information regulation 2, S.cerevisiae, homolog 7; Sir2l7; SIR2-like protein 7; sir2-related protein type 7; Sirt7; sirtuin 7; sirtuin type 7; Unknown (protein for MGC:142712) | |
| Mouse | |
| Protein G | |
| RUO | |
| 51547 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction