missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRT7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20375
This item is not returnable.
View return policy
Description
SIRT7 Polyclonal specifically detects SIRT7 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SIRT7 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| EC 3.5.1.-, MGC126840, MGC126842, silent mating type information regulation 2, S.cerevisiae, homolog 7, SIR2L7NAD-dependent deacetylase sirtuin-7, SIR2-like protein 7, sir2-related protein type 7, sirtuin (silent mating type information regulation 2 homolog) 7 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S.cerevisiae, homolog) 7, sirtuin 7, sirtuin type 7 | |
| Rabbit | |
| 45 kDa | |
| 0.1 mL | |
| DNA Repair, Epigenetics, Histone Deacetylases | |
| 51547 | |
| Mouse | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q9NRC8 | |
| SIRT7 | |
| Recombinant protein encompassing a sequence within the center region of human SIRT7. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Mouse (98%), Pig (94%), Rat (98%), Bovine (95%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction