missing translation for 'onlineSavingsMsg'
Learn More

SIRT7 Antibody (1E2G10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£173.00 - £435.00

Specifications

Antigen SIRT7
Clone 1E2G10
Concentration 1 mg/mL
Dilution Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18149939
View Documents
Novus Biologicals
NBP2-52565
0.1 mg
£435.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18629648
View Documents
Novus Biologicals
NBP2-52565-0.025mg
0.025 mg
£173.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SIRT7 Monoclonal specifically detects SIRT7 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications

Specifications

SIRT7
1 mg/mL
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF, Immunocytochemistry
Unconjugated
Mouse
DNA Repair, Epigenetics, Histone Deacetylases
PBS with 0.05% Sodium Azide
51547
Purified recombinant fragment of human SIRT7 (AA: 1-105) expressed in E. Coli.
Primary
44.9 kDa
1E2G10
Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready
Monoclonal
Purified
RUO
Human
EC 3.5.1.-, MGC126840, MGC126842, silent mating type information regulation 2, S.cerevisiae, homolog 7, SIR2L7NAD-dependent deacetylase sirtuin-7, SIR2-like protein 7, sir2-related protein type 7, sirtuin (silent mating type information regulation 2 homolog) 7 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S.cerevisiae, homolog) 7, sirtuin 7, sirtuin type 7
SIRT7
IgG1
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.