missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIRT6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118404
This item is not returnable.
View return policy
Description
SIRT6 Polyclonal Antibody for IHC (P), Flow
Sirtuins regulate epigenetic gene silencing. SIRT6 (Mono-ADP-ribosyltransferase sirtuin-6), a mouse Sir2 homolog, is a chromatin-associated nuclear ADP-ribosyltransferase protein. It appears to be involved in DNA repair and may also play a role in human aging.
Specifications
| SIRT6 | |
| Polyclonal | |
| Unconjugated | |
| SIRT6 | |
| 2810449N18Rik; AI043036; NAD-dependent deacetylase sirtuin-6; NAD-dependent protein deacetylase sirtuin-6; Regulatory protein SIR2 homolog 6; SIR2; SIR2 like 6; Sir2l6; SIR2-like protein 6; sir2-related protein type 6; Sirt6; sirtuin 6; sirtuin type 6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 50721, 51548 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P59941, Q8N6T7 | |
| SIRT6 | |
| Synthetic peptide within N-terminal human SIRT6. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction