missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SIRT6 Monoclonal Antibody (2G1H1)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA1214

Product Code. 15904314

  • £417.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA1-214 SIRT6 antibody detects SIRT6 by western blot and immunofluroescence. MA1-214 SIRT6 antibody detects SIRT6 by western blot at ~42kDa.

Sirtuins regulate epigenetic gene silencing. SIRT6 (Mono-ADP-ribosyltransferase sirtuin-6), a mouse Sir2 homolog, is a chromatin-associated nuclear ADP-ribosyltransferase protein. It appears to be involved in DNA repair and may also play a role in human aging.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SIRT6
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
Q8N6T7
SIRT6
Purified recombinant fragment of human SIRT6 (AA: 141-250) expressed in E. Coli.
100 μg
Primary
Human
Antibody
IgG1
Western Blot, Immunocytochemistry, Western Blot
2G1H1
Unconjugated
SIRT6
2810449N18Rik; AI043036; NAD-dependent deacetylase sirtuin-6; NAD-dependent protein deacetylase sirtuin-6; Regulatory protein SIR2 homolog 6; SIR2; SIR2 like 6; Sir2l6; SIR2-like protein 6; sir2-related protein type 6; Sirt6; sirtuin 6; sirtuin type 6
Mouse
Protein A
RUO
51548
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.