missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Listeria monocytogenes antigen 2 Antibody (2:13 C5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54650-0.025mg
This item is not returnable.
View return policy
Description
Listeria monocytogenes antigen 2 Monoclonal antibody specifically detects Listeria monocytogenes antigen 2 in Bacteria samples. It is validated for Western Blot, ELISA.
Specifications
| Listeria monocytogenes antigen 2 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000 | |
| EC 3.5.1.-, NAD-dependent deacetylase sirtuin-6, SIR2L6sir2-related protein type 6, SIR2-like protein 6, sirtuin (silent mating type information regulation 2 homolog) 6 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S. cerevisiae, homolog) 6, sirtuin 6, sirtuin type 6 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Specific for Listeria monocytogenes, not reactive for S. aureus, S. faecalis, B. subtilis, E. coli | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 2:13 C5 | |
| Unconjugated | |
| PBS with 0.5% Sodium Azide | |
| SIRT6 | |
| Listeria monocytogenes antigen 2 protein | |
| 0.025 mg | |
| Virology Bacteria and Parasites | |
| 51548 | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction