missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10099-1-AP-20UL
This item is not returnable.
View return policy
Description
SIRT3 is a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The SIRT3 is included in class I of the sirtuin family.Specifications
| SIRT3 | |
| Polyclonal | |
| Unconjugated | |
| SIRT3 | |
| hSIRT3, SIR2 like protein 3, SIR2L3, SIRT3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23410, 293615, 64384 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8R104, Q9NTG7 | |
| Sirt3 | |
| SIRT3 Fusion Protein Ag0148 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction