missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA125472
This item is not returnable.
View return policy
Description
Recommended positive controls: NIH3T3. Predicted molucular weight of 43 kDa.
The SIRT2 protein (also known as Sirtuin for Silent Mating Type Information 2-Homolog) is a NAD-dependent deacytylase (NDAC) that has been shown to control gene silencing, cell cycle, and DNA damage repair. It is believed that SIRT2 may act as a tumor suppressor in human gliomas and may also serve as a novel molecular marker for these cells. SIRT2 has also been shown to act as a redox sensor to help regulate muscle gene expression in response to food intake and exercise. SIRT2 acts in the phosphorylation cascade involving mitosis where SIRT2 is phosphorylated in late G2 phase, during M phase, and into cytokinesis.
Specifications
| SIRT2 | |
| Polyclonal | |
| Unconjugated | |
| SIRT2 | |
| 5730427M03Rik; 5E5 antigen; mSIR2L2; NAD-dependent deacetylase sirtuin-2; NAD-dependent protein deacetylase sirtuin-2; Regulatory protein SIR2 homolog 2; silent information regulator 2; silent mating type information regulation 2, (S.cerevisiae, homolog)-like; sirtuin 2; SIR2; SIR2L; SIR2L2; SIR2-like protein 2; sir2-related protein type 2; Sirt2; sirtuin 2; sirtuin type 2; sirtuin-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22933, 64383 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| 1.33 mg/mL | |
| 0.01M PBS with 1% BSA and 15mM sodium azide; pH 7.4 | |
| Q8IXJ6, Q8VDQ8 | |
| SIRT2 | |
| Synthetic peptide corresponding to the C-terminal residues 722-737 of mouse SIR2/SIRT2, with a C-terminal added lysine. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction