Learn More
SIRT1 Rabbit, anti-Human, CoraLite Plus 750, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | SIRT1 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Flow Cytometry (Intracellular Staining) |
| Conjugate | CoraLite Plus 750 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18030526
|
Proteintech
CL750-13161-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18014795
|
Proteintech
CL750-13161-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SIRT1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SIRT1. This antibody recognizes human antigen. The SIRT1 antibody has been validated for the following applications: FC (Intra), WB analysis. This antibody is coralite™ plus 750.
Specifications
| SIRT1 | |
| Western Blot, Flow Cytometry (Intracellular Staining) | |
| Liquid | |
| Human | |
| hSIR2, hSIRT1, SIR2 like protein 1, SIR2L1, SIRT1 | |
| Primary |
| 1000 μg/mL | |
| CoraLite Plus 750 | |
| Rabbit | |
| 23411 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.