missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SIRT1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA585921

Product Code. 16382825

  • £440.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SIRT1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

NAD-dependent protein deacetylase sirtuin-1 (SIRT1) links transcriptional regulation directly to intracellular energetics and participates in the coordination of several separated cellular functions such as cell cycle, repsonse to DNA damage, metabolism, apoptosis, and autophagy. SIRT1 can modulate chromatin function through deacetylation of histones and can promote alterations in the methylation of histone and DNA, leading to transcriptional repression. It is essential in skeletal muscle cell differentiation and in response to low nutrients mediates the inhibitory effect on skeletal myoblast differentiation which also involves 5'-AMP-activated protein kinase (AMPK) and nicotinamide phosphoribosyltransferase (NAMPT).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SIRT1
Polyclonal
Unconjugated
SIRT1
75SirT1; AA673258; hSIR2; hSIRT1; mSIR2a; NAD-dependent deacetylase sirtuin-1; NAD-dependent protein deacetylase sirtuin-1; OTTHUMP00000198111; OTTHUMP00000198112; Regulatory protein SIR2 homolog 1; RP11-57G10.3; SIR2; Sir2a; SIR2alpha; SIR2L1; sir2-like 1; SIR2-like protein 1; SIRT 1; SIRT1; SIRT-1; SirtT1 75 kDa fragment; sirtuin 1; sirtuin 1 ((silent mating type information regulation 2, homolog) 1; sirtuin type 1
Rabbit
Antigen affinity chromatography
RUO
23411, 93759
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Q923E4, Q96EB6
SIRT1
Synthetic peptide within Human SIRT1 aa 698-747.
100 μL
Primary
Human, Mouse
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Promozioni

Promozioni

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato