Learn More
SIRPB2 Antibody (1008702) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB99981I
Product Code. 18593949
  • £461.00 / 0.10mL
Description
SIRPB2 Monoclonal antibody specifically detects SIRPB2 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SIRPB2
Monoclonal
Janelia Fluor 549
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
284759
Store at 4°C in the dark.
IgG2b
Flow Cytometry, CyTOF
1008702
Flow Cytometry, CyTOF-ready
dJ776F14.2, Protein Tyrosine Phosphatase Non-Receptor Type Substrate 1-Like 3, Protein Tyrosine Phosphatase Non-Receptor Type Substrate Protein, Protein Tyrosine Phosphatase, Non-Receptor Type 1-Like, Protein Tyrosine Phosphatase, Non-Receptor Type Substrate 1-Like 3, PTPN1L, PTPNS1L3, Signal-Regulatory Protein Beta 2, Signal-Regulatory Protein Beta-2, SIRP-beta-2, SIRPG
HEK293 human embryonic kidney cell line, HEK293 derived Human SIRP beta 2, Gln33-Gly287, Accession # Q5JXA9
0.1 mL
Primary
Human
Purified