missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIRPB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA535262
This item is not returnable.
View return policy
Description
SIRPB1 Polyclonal Antibody for Western Blot, IHC (P)
SIRP beta 1 is a type I transmembrane protein belonging to the SIRP family within the Ig superfamily. Members of this family are characterized by an extracellular region containing a V-set Ig domain with a J-like sequence and two C1-set Ig domains. Unlike SIRP alpha that has cytoplasmic ITIM domains, SIRP beta 1 possesses positively charged residues that allow association with ITAM motif-containing adaptor molecules. SIRP beta 1 is expressed on cells of monocyte, macrophage or dendritic lineages.
Specifications
| SIRPB1 | |
| Polyclonal | |
| Unconjugated | |
| SIRPB1 | |
| 9930027N05Rik; CD172 antigen-like family member B; CD172b; CD172b antigen; DKFZp686A05192; signal regulatory protein beta 1; signal-regulatory protein beta 1; signal-regulatory protein beta 1A; signal-regulatory protein beta-1; Signal-regulatory protein beta-1 isoform 3; SIRP BETA 1; SIRP beta 1 cell surface protein; Sirpb; SIRPB1; Sirpb1a; SIRP-beta; SIRP-beta B; SIRP-BETA-1; SIRP-beta-1 isoform 3 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 10326 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.34 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O00241 | |
| SIRPB1 | |
| KLH conjugated synthetic peptide between 17-43 amino acids from the N-terminal region of human SIRPB1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction