missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIRPB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598148
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
SIRPB1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
SIRP beta 1 is a type I transmembrane protein belonging to the SIRP family within the Ig superfamily. Members of this family are characterized by an extracellular region containing a V-set Ig domain with a J-like sequence and two C1-set Ig domains. Unlike SIRP alpha that has cytoplasmic ITIM domains, SIRP beta 1 possesses positively charged residues that allow association with ITAM motif-containing adaptor molecules. SIRP beta 1 is expressed on cells of monocyte, macrophage or dendritic lineages.
Specifikationer
| SIRPB1 | |
| Polyclonal | |
| Unconjugated | |
| SIRPB1 | |
| 9930027N05Rik; CD172 antigen-like family member B; CD172b; CD172b antigen; DKFZp686A05192; signal regulatory protein beta 1; signal-regulatory protein beta 1; signal-regulatory protein beta 1A; signal-regulatory protein beta-1; Signal-regulatory protein beta-1 isoform 3; SIRP BETA 1; SIRP beta 1 cell surface protein; Sirpb; SIRPB1; Sirpb1a; SIRP-beta; SIRP-beta B; SIRP-BETA-1; SIRP-beta-1 isoform 3 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 10326 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2.13 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O00241 | |
| SIRPB1 | |
| Recombinant Human Signal-regulatory protein beta-1 protein (30-371AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering