missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIRP gamma Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547627
This item is not returnable.
View return policy
Description
In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) SIRP alpha, rhSIRP beta 1, and recombinant mouse SIRP alpha is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
SIRP gamma (CD172g) recognizes a 55 kDa signal regulatory protein which is a member of the immunoglobulin gene superfamily. SIRP gamma is expressed on most T lymphocytes and a subset of B-cells but is absent on myeloid cells. CD172g has a truncated cytoplasmic tail, which is similar to SIRP beta, but unlike SIRP beta CD172g does not require DAP12 for expression at the cell surface. CD172g, like SIRP alpha, binds to CD47 inducing apoptosis. SIRP gamma is also involved in the negative regulation of receptor tyrosine kinase-coupled signalling.
Specifications
| SIRP gamma | |
| Polyclonal | |
| Unconjugated | |
| SIRPG | |
| bA77C3.1; CD172 antigen-like family member B; CD172 gamma; CD172g; CD172g antigen; RP11-77C3.2; signal regulatory protein gamma; signal-regulatory protein beta 2; signal-regulatory protein beta-2; Signal-regulatory protein gamma; SIRP beta 2; SIRPB2; SIRP-B2; SIRP-beta-2; SIRPG; SIRPgamma; SIRP-gamma | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 55423 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9P1W8 | |
| SIRPG | |
| CHO-derived recombinant human SIRP gamma/CD172g Val64-Ser364, (Val263Ala) and (Ser286Leu). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction