missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SIRP gamma Monoclonal Antibody (OX-119)
GREENER_CHOICE

Product Code. 16322504
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16322504 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16322504 Supplier Invitrogen™ Supplier No. MA528217

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse anti Human CD172g antibody, clone OX119 recognizes human SIRP gamma, a 55 kDa signal regulatory protein which is a member of the immunoglobulin gene superfamily.

SIRP gamma (CD172g) recognizes a 55 kDa signal regulatory protein which is a member of the immunoglobulin gene superfamily. SIRP gamma is expressed on most T lymphocytes and a subset of B-cells but is absent on myeloid cells. CD172g has a truncated cytoplasmic tail, which is similar to SIRP beta, but unlike SIRP beta CD172g does not require DAP12 for expression at the cell surface. CD172g, like SIRP alpha, binds to CD47 inducing apoptosis. SIRP gamma is also involved in the negative regulation of receptor tyrosine kinase-coupled signalling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRP gamma
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone OX-119
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene SIRPG
Gene Accession No. Q9P1W8
Gene Alias bA77C3.1; CD172 antigen-like family member B; CD172 gamma; CD172g; CD172g antigen; RP11-77C3.2; signal regulatory protein gamma; signal-regulatory protein beta 2; signal-regulatory protein beta-2; Signal-regulatory protein gamma; SIRP beta 2; SIRPB2; SIRP-B2; SIRP-beta-2; SIRPG; SIRPgamma; SIRP-gamma
Gene Symbols SIRPG
Host Species Mouse
Immunogen Purified recombinant SIRP gamma CD4.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55423
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.