missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRP gamma/CD172g Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82342-0.1ML
This item is not returnable.
View return policy
Description
SIRP gamma/CD172g Polyclonal specifically detects SIRP gamma/CD172g in Human samples. It is validated for Western Blot.
Specifications
| SIRP gamma/CD172g | |
| Polyclonal | |
| PBS, 2% sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| bA77C3.1, CD172g, CD172g antigen, FLJ42230, signal-regulatory protein beta 2, Signal-regulatory protein beta-2, signal-regulatory protein gamma, SIRP beta 2, SIRP-B2, SIRPB2CD172 antigen-like family member B, SIRP-beta-2, SIRPgamma, SIRP-gamma | |
| The immunogen is a synthetic peptide directed towards the middle region of Human SIRP gamma | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 55423 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction