missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRP Beta 1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68206-1-IG-20UL
This item is not returnable.
View return policy
Description
The SIRP Beta 1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SIRP Beta 1. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The SIRP Beta 1 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| SIRP Beta 1 | |
| Monoclonal | |
| 1000 μg/mL | |
| CD172b; DKFZp686A05192; FLJ26614; SIRP-BETA-1 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat, Rabbit, Pig | |
| 43 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 3A3H1 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 10326 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction