missing translation for 'onlineSavingsMsg'
Learn More

SIRP beta 1/gamma Antibody (OX117), DyLight 350, Novus Biologicals™

Product Code. 18368690 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368690 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368690 Supplier Novus Biologicals Supplier No. NBP281071UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SIRP beta 1/gamma Monoclonal specifically detects SIRP beta 1/gamma in Human samples. It is validated for Flow Cytometry, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRP beta 1/gamma
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone OX117
Conjugate DyLight 350
Dilution Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Recombinant human SIRPgamma:CD4 fusion protein.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody binds to human SIRP beta 1/gamma.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.