missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRP beta 1/gamma Antibody (OX117), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-81071AF350
This item is not returnable.
View return policy
Description
SIRP beta 1/gamma Monoclonal specifically detects SIRP beta 1/gamma in Human samples. It is validated for Flow Cytometry, ELISA.
Specifications
| SIRP beta 1/gamma | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Recombinant human SIRPgamma:CD4 fusion protein. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| OX117 | |
| Flow Cytometry, ELISA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| This antibody binds to human SIRP beta 1/gamma. | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction