missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRP beta 1/CD172b Antibody (010), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90729AF405
This item is not returnable.
View return policy
Description
SIRP beta 1/CD172b Monoclonal specifically detects SIRP beta 1/CD172b in Mouse samples. It is validated for ELISA.
Specifications
| SIRP beta 1/CD172b | |
| Monoclonal | |
| Alexa Fluor 405 | |
| CD172 antigen-like family member B, CD172b, CD172b antigen, DKFZp686A05192, FLJ26614, signal-regulatory protein beta 1, signal-regulatory protein beta-1, SIRP-BETA-1, SIRP-beta-1 isoform 3 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse SIRP beta 1/CD172b (BAD26610.1; Met1-Lys363). | |
| 0.1 mL | |
| Signal Transduction | |
| 10326 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 10 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction