missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRP alpha/CD172a Antibody (009), Janelia Fluor™ 646, Novus Biologicals™
Shop All Bio Techne ProductsDescription
SIRP alpha/CD172a Monoclonal antibody specifically detects SIRP alpha/CD172a in Mouse samples. It is validated for ELISA
Specifications
Specifications
| Conjugate | Janelia Fluor 646 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Host Species | Rabbit |
| Research Discipline | GPCR, Protein Phosphatase, Signal Transduction |
| Content And Storage | Store at 4C in the dark. |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?