missing translation for 'onlineSavingsMsg'
Learn More

SIRP alpha/CD172a Antibody (001), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18340840 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18340840 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18340840 Supplier Novus Biologicals Supplier No. NBP290751AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SIRP alpha/CD172a Monoclonal specifically detects SIRP alpha/CD172a in Mouse samples. It is validated for Flow Cytometry, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRP alpha/CD172a
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 1
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias Bit, BITbrain-immunoglobulin-like molecule with tyrosine-based activation motifs, Brain Ig-like molecule with tyrosine-based activation motifs, CD172 antigen-like family member A, CD172a, CD172a antigen, Inhibitory receptor SHPS-1, Macrophage fusion receptor, MFRtyrosine phosphatase SHP substrate 1, MYD1, MYD-1, MyD-1 antigen, P84, protein tyrosine phosphatase, non-receptor type substrate 1, PTPNS1, SHP substrate 1, SHPS-1, SHPS1CD172A, signal-regulatory protein alpha, Signal-regulatory protein alpha-1, Signal-regulatory protein alpha-2, Signal-regulatory protein alpha-3, SIRPalpha, SIRP-ALPHA-1, Sirp-alpha-2, SIRPalpha2, Sirp-alpha-3, SIRPtyrosine-protein phosphatase non-receptor type substrate 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse SIRP alpha/CD172a (BAA20376.1; Met1-Asn373).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Protein Phosphatase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 140885
Test Specificity Has cross-reactivity in ELISA with: Human SIRPA
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.