missing translation for 'onlineSavingsMsg'
Learn More

Siremadlin (R Enantiomer), MedChemExpress

Product Code. 16413587
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16413587

Brand: Medchem Express HY18658A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Siremadlin R Enantiomer (NVP-HDM201 R Enantiomer) is the R enantiomer of Siremadlin. Siremadlin is a potent and highly specific MDM-2/p53 inhibitor.

Siremadlin R Enantiomer (NVP-HDM201 R Enantiomer) is the R enantiomer of Siremadlin. Siremadlin is a potent and highly specific MDM-2/p53 inhibitor.

Specifications

Chemical Name or Material Siremadlin (R Enantiomer)
CAS 1448867-42-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H24Cl2N6O4
Quantity 50 mg
Synonym NVP-HDM201 (R Enantiomer) HDM201 (R Enantiomer)
Solubility Information DMSO : ≥ 56.75 mg/mL (102.18 mM)
SMILES O=C1N(C2=CC(Cl)=CN(C)C2=O)[C@H](C3=CC=C(Cl)C=C3)C4=C1N=C(C5=CN=C(OC)N=C5OC)N4C(C)C
Molecular Weight (g/mol) 555.41
Formula Weight 555.41
Percent Purity 99.02%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.