missing translation for 'onlineSavingsMsg'
Learn More

Siremadlin, MedChemExpress

Product Code. 16497558
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16497558

Brand: Medchem Express HY18658

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Siremadlin (NVP-HDM201) is a potent, orally bioavailable and highly specific p53-MDM2 interaction inhibitor.

Siremadlin (NVP-HDM201) is a potent, orally bioavailable and highly specific p53-MDM2 interaction inhibitor.

Specifications

Chemical Name or Material Siremadlin
CAS 1448867-41-1
Color White
Purity Grade Notes Research
Molecular Formula C26H24Cl2N6O4
Quantity 10 mg
Synonym NVP-HDM201 HDM201
Solubility Information DMSO : ≥ 56.75 mg/mL (102.18 mM)
SMILES O=C1N(C2=CC(Cl)=CN(C)C2=O)[C@@H](C3=CC=C(Cl)C=C3)C4=C1N=C(C5=CN=C(OC)N=C5OC)N4C(C)C
Molecular Weight (g/mol) 555.41
Formula Weight 555.41
Percent Purity 99.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.