missing translation for 'onlineSavingsMsg'
Learn More

Siponimod, MedChemExpress

Product Code. 16487837
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
500mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487837 5 mg 5mg
16471239 1 g 1g
16465118 10 mg 10mg
16431357 50 mg 50mg
16411636 500 mg 500mg
16405846 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16487837 Supplier Medchem Express Supplier No. HY12355
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Siponimod (BAF-312) is an orally active and selective sphingosine-1-phosphate (S1P) receptor modulator. Siponimod is selective for S1P1 and S1P5 over S1P2, S1P3, and S1P4, with EC50s of 0.4, 0.98, >10000, >1000, and 750 nM, respectively. Siponimod can be used for multiple sclerosis (MS) research-.

Siponimod (BAF-312) is an orally active and selective sphingosine-1-phosphate (S1P) receptor modulator. Siponimod is selective for S1P1 and S1P5 over S1P2, S1P3, and S1P4, with EC50s of 0.4, 0.98, >10000, >1000, and 750 nM, respectively. Siponimod can be used for multiple sclerosis (MS) research-.

Specifications

Chemical Name or Material Siponimod
CAS 1230487-00-9
Color White
Purity Grade Notes Research
Molecular Formula C29H35F3N2O3
Quantity 5 mg
Synonym BAF-312
Solubility Information DMSO : ≥ 30 mg/mL (58.07 mM)
SMILES O=C(C1CN(CC2=CC=C(/C(C)=N/OCC3=CC=C(C4CCCCC4)C(C(F)(F)F)=C3)C=C2CC)C1)O
Molecular Weight (g/mol) 516.6
Formula Weight 516.6
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.