missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Sipatrigine
GREENER_CHOICE

Product Code. 18085642
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18085642

Brand: Tocris Bioscience™ 4044/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

NaV blocker; neuroprotective

Blocker of voltage-dependent sodium channels (NaV). Inhibits glutamate release; displays neuroprotective activity in rat models of cerebral ischemia. Also thought to block Ca2+ channels. Analog of lamotrigine (Cat. No. 1611).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130800-90-7
Molecular Formula C15H16Cl3N5

Specifications

Quantity 10 mg
Formula Weight 372.68
Purity >99%
Molecular Weight (g/mol) 372.678
Chemical Name or Material Sipatrigine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.