missing translation for 'onlineSavingsMsg'
Learn More

Sipatrigine, MedChemExpress

Product Code. 16416907
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 16416907

Brand: Medchem Express HY108335

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sipatrigine (619C89), a neuroprotective agent, is a glutamate release inhibitor, voltage-dependent sodium channel and calcium channel inhibitor, penetrating the central nervous system. Has the potential in the study for focal cerebral ischemia and stroke.

Sipatrigine (619C89), a neuroprotective agent, is a glutamate release inhibitor, voltage-dependent sodium channel and calcium channel inhibitor, penetrating the central nervous system. Has the potential in the study for focal cerebral ischemia and stroke.

Specifications

Chemical Name or Material Sipatrigine
CAS 130800-90-7
Color White
Purity Grade Notes Research
Molecular Formula C15H16Cl3N5
Quantity 5 mg
Synonym 619C89 BW 619C89
Solubility Information DMSO : 50 mg/mL (134.16 mM; Need ultrasonic)
SMILES NC1=NC(N2CCN(C)CC2)=NC=C1C3=CC(Cl)=CC(Cl)=C3Cl
Molecular Weight (g/mol) 372.68
Formula Weight 372.68
Percent Purity 99.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.