missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SIPA1L3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12848972
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12848972 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12848972 Supplier Invitrogen™ Supplier No. PA520849

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is SK-N-SH cell lysate. PA5-20849 can be used with blocking peptide PEP-0963.

Signal-induced proliferation associated-like protein 2 (SIPA1L3) is a member of the SIPA1 family of RapGAPs. SIPA1L3 was cloned as a novel molecular component of tight junctions and adherens junctions, suggesting that may function to balance Rap1 by counteracting RapGEFs. Recent studies of SIPA indicate that its deregulation can cause myeloproliferative stem cell disorders in mice and increased metastases in human cancers. Other studies suggest SIPA1L1 may play important roles in embryo development and control of cell proliferation. Based on the amount of homology between SIPA family members, it is possible that SIPA1L3 plays other roles development and cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIPA1L3
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SIPA1L3
Gene Accession No. G3X9J0, O60292
Gene Alias 2610511M17Rik; CTRCT45; KIAA0545; signal induced proliferation associated 1 like 3; signal-induced proliferation-associated 1 like 3; signal-induced proliferation-associated 1-like protein 3; Sipa1l3; SIPA1-like protein 3; SPA-1-like 3; SPA-1-like protein 3; Spal3; SPAR3
Gene Symbols SIPA1L3
Host Species Rabbit
Immunogen A 15 amino acid peptide from near the amino terminus of human SIPA1L3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23094, 292771, 74206
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.