missing translation for 'onlineSavingsMsg'
Learn More

SIPA1L3 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18400656 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400656 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400656 Supplier Novus Biologicals Supplier No. NBP230362AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SIPA1L3 Polyclonal antibody specifically detects SIPA1L3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIPA1L3
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias KIAA0545, Signal-Induced Proliferation-Associated 1 Like 3, Signal-Induced Proliferation-Associated 1-Like Protein 3, SIPA1-Like Protein 3, SPA-1-Like 3, SPA-1-Like Protein 3, SPAL3
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1 to 50 of human Signal-Induced Proliferation-Associated 1 Like 3 using the numbering given in entry NP_055888.1 (GeneID 23094).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 23094
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.