missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SIPA1L2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12828972
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12828972 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12828972 Supplier Invitrogen™ Supplier No. PA520848

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is rat brain tissue lysate. PA5-20848 can be used with blocking peptide PEP-0962.

Signal-induced proliferation associated-like protein 2 (SIPA1L2) is a member of the SIPA1 family of RapGAPs. Little is known of the role of the SIPA1L2 protein, but recent studies of SIPA indicate that its deregulation can cause myeloproliferative stem cell disorders in mice and increased metastases in human cancers. Other studies suggest SIPA1L1 may play important roles in embryo development and control of cell proliferation. Based on the amount of homology between SIPA family members, it is likely that SIPA1L2 plays a role in embryo development and cell proliferation, possibly including oncogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIPA1L2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SIPA1L2
Gene Accession No. Q5JCS6, Q80TE4, Q9P2F8
Gene Alias BC058408; Kiaa0545; KIAA1389; mKIAA1389; serine-rich synapse associated protein 2; serine-rich synapse-associated protein; Sersap2; signal induced proliferation associated 1 like 2; signal-induced proliferation-associated 1 like 2; signal-induced proliferation-associated 1-like protein 2; Sipa1l2; SIPA1-like protein 2; SPA-1-like 2; SPAL2; Spar2
Gene Symbols SIPA1L2
Host Species Rabbit
Immunogen A 19 amino acid peptide from near the amino terminus of human SIPA1L2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 244668, 361442, 57568
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.