missing translation for 'onlineSavingsMsg'
Learn More

SIPA1L1 Polyclonal Antibody

Product Code. 12818972
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12818972 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12818972 Supplier Invitrogen Supplier No. PA520847

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SIPA1L1 Polyclonal Antibody

SIPA1L1 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen SIPA1L1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SIPA1L1
Gene Accession No. O35412, O43166, Q8C0T5
Gene Alias 4931426N11Rik; AW213287; E6-targeted protein 1; E6TP1; H_DJ1140G11.1; High-risk human papilloma viruses E6 oncoproteins targeted protein 1; Kiaa0440; mKIAA0440; signal induced proliferation associated 1 like 1; signal-induced proliferation-associated 1 like 1; signal-induced proliferation-associated 1-like protein 1; SIPA1L1; SIPA1-like protein 1; Spa1; Spa-1; SPA-1 like protein p1294; SPA-1-like protein p1294; SPAL; Spar; Spine-associated Rap GTPase-activating protein; spine-associated Rap guanosine triphosphatase (GTPase) activating protein (GAP); WUGSC:H_DJ1140G11.1
Gene Symbols SIPA1L1
Host Species Rabbit
Immunogen A 16 amino acid peptide from near the amino terminus of human SIPA1L1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217692, 246212, 26037
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.