missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIPA1 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA518245
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
The product of this gene is a mitogen induced GTPase activating protein. It exhibits a specific GAP activity for Ras-related regulatory proteins Rap1 and Rap2, but not for Ran or other small GTPases. This protein may also hamper mitogen-induced cell cycle progression when abnormally or prematurely expressed. It is localized to the perinuclear region. Two alternatively spliced variants encoding the same isoform have been characterized to date.
Tekniske data
| SIPA1 | |
| Polyclonal | |
| Unconjugated | |
| SIPA1 | |
| GTPase-activating protein Spa-1; MGC102688; MGC17037; p130 SPA-1; signal-induced proliferation associated gene 1; signal-induced proliferation-associated 1; signal-induced proliferation-associated gene 1; signal-induced proliferation-associated protein 1; SIPA1; Sipa-1; SPA1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 6494 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q96FS4 | |
| SIPA1 | |
| Synthetic peptide sequence (ASKQLGSPTADLA) corresponding to the C-terminus amino acids of SIPA1 (aa 1030-1042). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion