missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIPA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35235-20ul
This item is not returnable.
View return policy
Description
SIPA1 Polyclonal antibody specifically detects SIPA1 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| SIPA1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| GTPase-activating protein Spa-1, MGC102688, MGC17037, p130 SPA-1, signal-induced proliferation-associated 1, signal-induced proliferation-associated protein 1, sipa-1, SPA1signal-induced proliferation-associated gene 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 763-1042 of human SIPA1 (NP_694985.29).,, Sequence:, ESGRPRRSFSELYTLSLQEPSRRGAPDPVQDEVQGVTLLPTTKQLLHLCLQDGGSPPGPGDLAEERTEFLHSQNSLSPRSSLSDEAPVLPNTTPDLLLATTAKPSVPSADSETPLTQDRPGSPSGSEDKGNPAPELRASFLPRTLSLRNSISRIMSEAGSGTLEDEWQAISEIASTCNTILESLSREGQPIPESGDPKGTPKSDAEPEPGNLSEKVSHLESMLRKLQEDLQKEKADRAALEEEVRSLRHNNRRLQAESESAATRLLLASKQLGSPTADLA | |
| 20 μL | |
| Apoptosis, Breast Cancer | |
| 6494 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction