missing translation for 'onlineSavingsMsg'
Learn More

SIPA1 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18413335 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413335 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413335 Supplier Novus Biologicals Supplier No. NBP281850UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SIPA1 Polyclonal antibody specifically detects SIPA1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIPA1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias GTPase-activating protein Spa-1, MGC102688, MGC17037, p130 SPA-1, signal-induced proliferation-associated 1, signal-induced proliferation-associated protein 1, sipa-1, SPA1signal-induced proliferation-associated gene 1
Host Species Rabbit
Immunogen SIPA1 antibody was raised against a 19 amino acid synthetic peptide from near the amino terminus of human SIPA1. The immunogen is located within amino acids 40 - 90 of SIPA1. Amino Acid Squence: DAGEARPPTPASPRARAHS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6494
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.