missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIPA1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-18917CL1
This item is not returnable.
View return policy
Description
SIPA1 Polyclonal antibody specifically detects SIPA1 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| SIPA1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| GTPase-activating protein Spa-1, MGC102688, MGC17037, p130 SPA-1, signal-induced proliferation-associated 1, signal-induced proliferation-associated protein 1, sipa-1, SPA1signal-induced proliferation-associated gene 1 | |
| The immunogen recognized by this antibody maps to a region between residue 992 to 1042 of human signal-induced proliferation-associated protein 1 using the numbering given in entry NP_006738.3 (GeneID 6494). | |
| 0.1 mL | |
| Apoptosis, Breast Cancer | |
| 6494 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction