missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21672-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes one of the proteins found in the SMN complex, which consists of several gemin proteins and the protein known as the survival of motor neuron protein. The SMN complex is localized to a subnuclear compartment called gems (gemini of coiled bodies) and is required for assembly of spliceosomal snRNPs and for pre-mRNA splicing. This protein interacts directly with the survival of motor neuron protein and it is required for formation of the SMN complex.Specifications
| SIP1 | |
| Polyclonal | |
| Unconjugated | |
| GEMIN2 | |
| Component of gems 2, Gemin 2, GEMIN2, SIP1, SIP1 delta, SMN interacting protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 66603, 84404, 8487 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O14893, Q9CQQ4, Q9QZP1 | |
| GEMIN2 | |
| SIP1 Fusion Protein Ag16500 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction