missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sinomenine hydrochloride, MedChemExpress
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Sinomenine hydrochloride (Cucoline hydrochloride), an alkaloid extracted from Sinomenium acutum, is a blocker of the NF-κB activation. Sinomenine also is an activator of μ-opioid receptor.
Specifications
Specifications
| Chemical Name or Material | Sinomenine hydrochloride |
| CAS | 6080-33-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C19H24ClNO4 |
| Quantity | 100 mg |
| Synonym | Cucoline hydrochloride |
| Solubility Information | DMSO : 100 mg/mL (273.34 mM; Need ultrasonic and warming) ∣H2O : 16.67 mg/mL (45.57 mM; Need ultrasonic) |
| SMILES | OC(C1=C2C[C@@]3([H])[C@](C=C4OC)([H])[C@@]1(CCN3C)CC4=O)=C(C=C2)OC.Cl |
| Molecular Weight (g/mol) | 365.85 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction