missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Sinomenine (Hydrochloride)
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
Description
- Sinomenine is a natural plant alkaloid commonly used to alleviate inflammation associated with rheumatoid arthritis (Wang and Li)
- It impairs signaling through Nuclear factor-kappaB (NF-κB; Sun et al.; Wang and Li) and enhances the bioavailability of some compounds, at least in part through an inhibition of drug export by transporters like P-glycoprotein (Kesarwani et al.; Liu et al.)
- Promotes self-renewal in cultured human and mouse embryonic stem cells (Desbordes et al.)
Specifications
Specifications
| CAS | 6080-33-7 |
| Synonym | Cocculine; Cucoline; NSC 76021 |
| Quantity | 50 mg |
| Molecular Formula | C19H23NO4 · HCl |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction