missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Sino Biological™ RNASET2 Recombinant Human Protein, His Tag
Description
Ribonuclease T2 (RNASET2), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
- Activity: The activity of this protein has not been tested
- Formulation: 20mM Tris, 500mM NaCl, 10% Glycerol, pH 7.4
- Reconstitution: No reconstitution is needed
This protein is manufactured by Sino Biological Inc.
Specifications
Specifications
| For Use With (Application) | Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories |
| Gene ID (Entrez) | 8635 |
| Molecular Weight (g/mol) | 28.5kDa |
| Purification Method | PAGE |
| Quantity | 5 μg |
| Storage Requirements | Store at -80 °C. Store aliquots under sterile conditions at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. |
| Structural Form | Full Length/Recombinant |
| Endotoxin Concentration | <1 EU/μg |
| Gene Alias | RP11-514O12.3, RNASE6PL, bA514O12.3 |
| Common Name | Ribonuclease T2 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?