Learn More
PVR (CD155) Recombinant Mouse Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 345) of mouse PVR precursor (NP_081790.1) precursor fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus
Brand: Invitrogen 50259M03H25
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Poliovirus receptor (PVR/CD155), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.- Activity: Measured by its binding ability in a functional ELISA; immobilized mouse DNAM1 at 2μg/mL (100μL/well) can bind mouse PVR with a linear range of 6.4-160ng/mL
- Formulation: Lyophilized in 50mM Tris, 10mM NaCl, pH 7.5, 100mM Glycine, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
63kDa | |
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
<1 EU/μg | |
Poliovirus receptor | |
His Tag/IgG1 Fc Tag | |
CD Proteins | |
Viral Host Cell Receptors |
52118 | |
5 x 5μg | |
Truncated/Recombinant/Cell Surface Protein | |
3830421F03Rik, CD155, D7Ertd458e, HVED, PVS, Taa1, Tage4, mE4 | |
PVR | |
Mammalian | |
Lyophilized | |
≥85% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.