missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD1LG2 Recombinant Human Protein His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 219) of human PDCD1LG2 (NP_079515.2) fused to a polyhistidine tag at the C-terminus
Brand: Invitrogen 10292H08H250
This item is not returnable.
View return policy
Description
Programmed cell death 1 ligand 2 (PDCD1LG2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human PDCD1LG2/PDL2 at 1μg/mL (100μL/well) can bind recombinant human PD1 with a linear range of 7.8-1000ng/mL
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
24kDa | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
PDCD1LG2, B7DC, Btdc, PDL2, CD273, PD-L2, PDCD1L2, mgC142238, mgC142240, bA574F11.2 | |
PDCD1LG2 |
5 x 50μg | |
<1 EU/μg | |
Programmed cell death 1 ligand 2 |
For Research Use Only. Not for use in diagnostic procedures.