missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARP1 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from the amino acids corresponding to the full length of human PARP1 (NP_001609.2) (Met 1-Trp 1014) fused to a polyhistidine tag at the C-terminus
£418.00 - £662.00
Specifications
For Use With (Application) | Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories |
---|---|
Molecular Weight (g/mol) | 114.5kDa |
Storage Requirements | Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. |
Endotoxin Concentration | <1 EU/μg |
Gene Alias | PARP1, RP11-125A15.2, ADPRT, ADPRT1, PARP, PARP-1, PPOL, pADPRT-1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15227615
|
Invitrogen
11040H08B25 |
5 x 5μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15267615
|
Invitrogen
11040H08B50 |
50μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Poly (Adp-Ribose) Polymerase 1 (PARP1), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: Measured by its binding ability in a functional ELISA; immobilized human PARP1 at 10μg/mL (100μL/well) can bind to biotinylated human HSP70, the EC50 of biotinylated human HSP70 is 0.035μg/mL
- Formulation: 50mM Tris, 100mM NaCl, pH 8, 2mM EDTA, 2mM PMSF, 0.5mM TCEP, 10% glycerol
- Reconstitution: No reconstitution needed
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
PARP1, RP11-125A15.2, ADPRT, ADPRT1, PARP, PARP-1, PPOL, pADPRT-1 | |
PARP1 | |
Insect | |
Other Proteins |
114.5kDa | |
<1 EU/μg | |
Poly (ADP-ribose) polymerase 1 | |
His Tag | |
Frozen | |
≥90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.