missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LGALS8 Recombinant Human Protein, N-GST Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding human LGALS8 (AAF19370.1) (Met 1-Trp 317) fused to a GST tag at the N-terminus
Brand: Invitrogen 10301H09E50
This item is not returnable.
View return policy
Description
Lectin, galactoside-binding, soluble, 8 (LGALS8), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.- Activity: Measured by its ability to agglutinate human red blood cells, the ED50 for this effect is typically 0.2-1μg/mL
- Formulation: Lyophilized in 50mM Tris, 0.1M NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
62.7kDa | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
Lectin, galactoside-binding, soluble, 8 |
50μg | |
LGALS8, RP11-385F5.1, Gal-8, PCTA-1, PCTA1, Po66-CBP | |
LGALS8 |
For Research Use Only. Not for use in diagnostic procedures.