missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL37 Recombinant Human Protein, Invitrogen™ Sino Biological™
Click to view available options
Quantity:
5 x 5 μg
Unit Size:
25µg
Description
Interleukin 37 (IL37), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
- Activity: The activity of this protein has not been tested
- Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Specifications
| For Use With (Application) | Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories |
| Gene ID (Entrez) | 27178 |
| Molecular Weight (g/mol) | 18.7kDa |
| Purification Method | PAGE |
| Quantity | 5 x 5 μg |
| Storage Requirements | Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. |
| Structural Form | Full Length/Recombinant |
| Endotoxin Concentration | <1 EU/μg |
| Gene Alias | IL37, FIL1, FIL1(ZETA), FIL1Z, IL-1F7, IL-1H, IL-1H4, IL-1RP1, IL-37, IL1F7, IL1H4, IL1RP1 |
| Common Name | Interleukin 37 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction