Learn More
IL1RAPL2 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 356) of human IL1RAPL2 (NP_059112.1) fused to a polyhistidine tag at the C-terminus
Brand: Invitrogen 10156H08H5
Code : NEW
Additional Details : Weight : 0.10000kg
Description
Interleukin 1 receptor accessory protein-like 2 (IL1RAPL2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.- Activity: Measured by its ability to inhibit IL1B dependent proliferation in D10.G4.1 mouse helper T cells; approximately 5-20μg/mL will inhibit 50% of the biological response due to 40pg/mL of IL1B
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
5μg | |
<1 EU/μg | |
Interleukin 1 receptor accessory protein-like 4 | |
His Tag | |
Cytokines & Receptors | |
NFκB Pathway Proteins, Cytokine Receptors |
40.7kDa | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
IL1R9, IL-1R9, TIGIRR-1, IL1RAPL-2 | |
IL1RAPL2 | |
Mammalian | |
Lyophilized | |
≥95% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.