missing translation for 'onlineSavingsMsg'
Learn More

IL1B Recombinant Human Protein, Invitrogen™ Sino Biological™

Product Code. p-5232780
Click to view available options
Quantity:
5 x 5 μg
50 μg
Unit Size:
25µg
50µg
This item is not returnable. View return policy

Product Code. 15224725

Brand: Invitrogen 10139HNAE25

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IL1B (NP_000567.1) (Ala 117-Ser 269)

Interleukin 1, beta (IL1B), a recombinant human protein, is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as immunogens, protein standards, and in other cell biology research applications.
  • Activity: 1.Measured by its binding ability in a functional ELISA; a. Immobilized recombinant human IL1B at 2μg/mL (100μL/well) can bind recombinant human IL1R2 with a linear range of 6.25-400ng/mL; b. Immobilized human IL1B at 10μg/mL (100μL/well) can bind human IL1R1, the EC50 of human IL1R1 is 0.48μg/mL; 2. Measured in a cell proliferation assay using D10.G4.1 mouse helper T cells, the ED50 for this effect is typically 0.5-5pg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
Gene ID (Entrez) 3553
Molecular Weight (g/mol) 17kDa
Quantity 5 x 5 μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Alias IL-1, IL1-BETA, IL1F2, IL1B
Common Name Interleukin 1, beta
Gene Symbol IL1B
Protein Tag Untagged
Expression System E. coli
Protein Family Cytokines & Receptors
Form Lyophilized
Purity or Quality Grade ≥90% by SDS-PAGE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.