missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL1A Recombinant Mouse Protein, Invitrogen™ Sino Biological™
Click to view available options
:
5ug
Description
Includes
One vial of lyophilized protein
Interleukin 1 alpha (IL1A), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
- Activity: 1. Measured in a cell proliferation assay using D10.G4.1 mouse helper T cells, the ED50 for this effect is typically 2-10pg/mL; 2. Measured by its binding ability in a functional ELISA; immobilized mouse IL1A at 10μg/mL (100μL/well) can bind mouse IL1R1, the EC50 of mouse IL1R1 is 0.13μg/mL
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Specifications
| For Use With (Application) | Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories |
| Molecular Weight (g/mol) | 17kDa |
| Quantity | 5 μg |
| Storage Requirements | Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. |
| Gene Alias | RP23-160G19.8, Il-1a |
| Common Name | Interleukin 1 alpha |
| Gene Symbol | IL1A |
| Protein Tag | Untagged |
| Expression System | E. coli |
| Protein Family | Cytokines & Receptors |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction