Learn More
IL13 Recombinant Human Protein, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding human IL-13 (AAK53823.1) (Met 1 - Asn 132)
Brand: Invitrogen 10369HNAC5
Code : NEW
Additional Details : Weight : 0.10000kg
Description
Interleukin 13 (IL13), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.- Activity: Measured in a cell proliferation assay using TF1 human erythroleukemic cells (Kitamura, T. et al. 1989, J. Cell Physiol. 140:323), the ED50 for this effect is typically 0.75-3.5ng/mL
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
5μg | |
<1 EU/μg | |
Interleukin 13 | |
Untagged | |
Cytokines & Receptors | |
Interleukins |
12.5kDa | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
ALRH, BHR1, IL-13, mgC116786, mgC116788, mgC116789, P600 | |
IL13 | |
Mammalian | |
Lyophilized | |
≥90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.